N2-[3-(Dimethylamino)propyl]-2,3-pyridinediamine hydrochloride

Supplier: Matrix Scientific

053258-500MG 053258-2.5G
101902-918EA 364.62 USD
101902-918 101902-920
N2-[3-(Dimethylamino)propyl]-2,3-pyridinediamine hydrochloride
N2-[3-(Dimethylamino)propyl]-2,3-pyridinediamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₉ClN₄
MDL Number: MFCD13562666
CAS Number: 1220029-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR