2-{[3-(Dimethylamino)propyl]amino}nicotinic acid hydrochloride

Supplier: Matrix Scientific

053257-500MG 053257-2.5G
101902-914EA 364.62 USD
101902-914 101902-916
2-{[3-(Dimethylamino)propyl]amino}nicotinic acid hydrochloride
2-{[3-(Dimethylamino)propyl]amino}nicotinic acid hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClN₃O₂
MDL Number: MFCD13562665
CAS Number: 1220017-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR