2-{[3-(Dimethylamino)propyl]amino}isonicotinic acid hydrochloride

Supplier: Matrix Scientific

053256-500MG 053256-2.5G
101902-910EA 364.62 USD
101902-910 101902-912
2-{[3-(Dimethylamino)propyl]amino}isonicotinic acid hydrochloride
2-{[3-(Dimethylamino)propyl]amino}isonicotinic acid hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClN₃O₂
MDL Number: MFCD13562664
CAS Number: 1220036-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR