N1-(6-Chloro-2-pyrazinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride

Supplier: Matrix Scientific

053255-500MG 053255-2.5G
101902-906EA 364.62 USD
101902-906 101902-908
N1-(6-Chloro-2-pyrazinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride
N1-(6-Chloro-2-pyrazinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₆Cl₂N₄
MW: 251.16 g/mol
MDL Number: MFCD13562663
CAS Number: 1220017-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR