N1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N3,N3-dimethyl-1,3-propanediamine hydrochloride

Supplier: Matrix Scientific

053254-500MG 053254-2.5G
101902-902EA 364.62 USD
101902-902 101902-904
N1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N3,N3-dimethyl-1,3-propanediamine hydrochloride
N1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-N3,N3-dimethyl-1,3-propanediamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆Cl₂F₃N₃
MW: 318.17 g/mol
MDL Number: MFCD13562662
CAS Number: 1220037-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR