N1-(6-Chloro-4-pyrimidinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride
Supplier: Matrix Scientific
053253-500MG
053253-2.5G
101902-898EA
364.62
USD
101902-898
101902-900
N1-(6-Chloro-4-pyrimidinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride
N1-(6-Chloro-4-pyrimidinyl)-N3,N3-dimethyl-1,3-propanediamine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₆Cl₂N₄ |
MDL Number:
MFCD13562661 CAS Number: 1220029-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...