N-Cyclohexyl-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053247-500MG
053247-2.5G
101902-874EA
364.62
USD
101902-874
101902-876
N-Cyclohexyl-4-piperidinecarboxamide hydrochloride
N-Cyclohexyl-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₃ClN₂O |
MDL Number:
MFCD08456915 CAS Number: 63214-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...