N-Cyclohexyl-3-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053246-500MG 053246-2.5G
101902-870EA 364.62 USD
101902-870 101902-872
N-Cyclohexyl-3-piperidinecarboxamide hydrochloride
N-Cyclohexyl-3-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₃ClN₂O
MDL Number: MFCD13562656
CAS Number: 937724-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR