N-Cyclohexyl-2-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053245-500MG
053245-2.5G
101902-866EA
364.62
USD
101902-866
101902-868
N-Cyclohexyl-2-piperidinecarboxamide hydrochloride
N-Cyclohexyl-2-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₂₃ClN₂O |
MDL Number:
MFCD13562655 CAS Number: 1236256-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...