N-Cyclohexyl-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053244-500MG 053244-2.5G
101902-862EA 364.62 USD
101902-862 101902-864
N-Cyclohexyl-2-pyrrolidinecarboxamide hydrochloride
N-Cyclohexyl-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O
MDL Number: MFCD13562654
CAS Number: 1236261-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR