2-Amino-N-cyclohexyl-3-phenylpropanamide hydrochloride
Supplier: Matrix Scientific
053243-500MG
053243-2.5G
101902-858EA
364.62
USD
101902-858
101902-860
2-Amino-N-cyclohexyl-3-phenylpropanamide hydrochloride
2-Amino-N-cyclohexyl-3-phenylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₃ClN₂O |
MDL Number:
MFCD13562653 CAS Number: 68398-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...