2-Amino-N-cyclohexyl-2-methylpropanamide hydrochloride
Supplier: Matrix Scientific
053242-500MG
053242-2.5G
101902-854EA
364.62
USD
101902-854
101902-856
2-Amino-N-cyclohexyl-2-methylpropanamide hydrochloride
2-Amino-N-cyclohexyl-2-methylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O |
MDL Number:
MFCD13562652 CAS Number: 1220029-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...