2-Amino-N-cyclohexyl-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

053241-500MG 053241-2.5G
101902-850EA 364.62 USD
101902-850 101902-852
2-Amino-N-cyclohexyl-3-methylbutanamide hydrochloride
2-Amino-N-cyclohexyl-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃ClN₂O
MDL Number: MFCD13562651
CAS Number: 1236254-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR