3-Amino-N-cyclohexylpropanamide hydrochloride

Supplier: Matrix Scientific

053240-500MG 053240-2.5G
101902-846EA 364.62 USD
101902-846 101902-848
3-Amino-N-cyclohexylpropanamide hydrochloride
3-Amino-N-cyclohexylpropanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClN₂O
MDL Number: MFCD13562650
CAS Number: 115022-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR