6-Chloro-N-cyclohexyl-2-pyridinamine

Supplier: Matrix Scientific

053236-500MG 053236-2.5G
101902-832EA 364.62 USD
101902-832 101902-834
6-Chloro-N-cyclohexyl-2-pyridinamine
6-Chloro-N-cyclohexyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂
MDL Number: MFCD13562647
CAS Number: 1220036-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR