5-Bromo-2-(cyclohexylamino)pyridine

Supplier: Matrix Scientific

053235-500MG 053235-2.5G
101902-828EA 364.62 USD
101902-828 101902-830
5-Bromo-2-(cyclohexylamino)pyridine
5-Bromo-2-(cyclohexylamino)pyridine

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂
MDL Number: MFCD11124008
CAS Number: 942050-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR