5-Bromo-N-cyclohexyl-3-methyl-2-pyridinamine

Supplier: Matrix Scientific

053234-500MG 053234-2.5G
101902-824EA 364.62 USD
101902-824 101902-826
5-Bromo-N-cyclohexyl-3-methyl-2-pyridinamine
5-Bromo-N-cyclohexyl-3-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂
MDL Number: MFCD13562646
CAS Number: 1219976-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR