2-Chloro-N-cyclohexylnicotinamide
Supplier: Matrix Scientific
Synonyms:
2-Chloro-N-cyclohexylpyridine-3-carboxamide
053233-500MG
053233-2.5G
101902-820EA
364.62
USD
101902-820
101902-822
2-Chloro-N-cyclohexylnicotinamide
2-Chloro-N-cyclohexylnicotinamide
Hazard: Irritant
Formula:
C₁₂H₁₅ClN₂O MW: 238.72 g/mol |
MDL Number:
MFCD01344059 CAS Number: 57841-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...