N2-Cyclohexyl-2,3-pyridinediamine

Supplier: Matrix Scientific

053232-500MG 053232-2.5G
101902-816EA 364.62 USD
101902-816 101902-818
N2-Cyclohexyl-2,3-pyridinediamine
N2-Cyclohexyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MDL Number: MFCD08692167
CAS Number: 41082-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR