2-(Cyclohexylamino)nicotinic acid

Supplier: Matrix Scientific

053231-500MG 053231-2.5G
101902-812EA 364.62 USD
101902-812 101902-814
2-(Cyclohexylamino)nicotinic acid
2-(Cyclohexylamino)nicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD01711773
CAS Number: 35812-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR