3-Chloro-N-cyclohexyl-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

053228-500MG 053228-2.5G
101902-800EA 364.62 USD
101902-800 101902-802
3-Chloro-N-cyclohexyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-cyclohexyl-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₄ClF₃N₂
MDL Number: MFCD11120883
CAS Number: 1041565-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR