6-Chloro-N-cyclohexyl-4-pyrimidinamine

Supplier: Matrix Scientific

Synonyms: 4-Chloro-6-(cyclohexylamino)pyrimidine

053227-500MG 053227-2.5G
101902-796EA 364.62 USD
101902-796 101902-798
6-Chloro-N-cyclohexyl-4-pyrimidinamine
6-Chloro-N-cyclohexyl-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MW: 211.7 g/mol
MDL Number: MFCD03669655
CAS Number: 61667-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR