4-(Cyclohexylamino)-3-nitrobenzoic acid
Supplier: Matrix Scientific
053225-500MG
053225-2.5G
101902-788EA
364.62
USD
101902-788
101902-790
4-(Cyclohexylamino)-3-nitrobenzoic acid
4-(Cyclohexylamino)-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₃H₁₆N₂O₄ |
MDL Number:
MFCD01156576 CAS Number: 333340-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...