Ethyl-3-amino-4-(cyclohexylamino)benzoate

Supplier: Matrix Scientific

Synonyms: Ferrostatin-1, Ethyl 3-amino-4-(cyclohexylamino)benzoate

053224-500MG 053224-2.5G
101902-784EA 330.77 USD
101902-784 101902-786
Ethyl-3-amino-4-(cyclohexylamino)benzoate
Ethyl-3-amino-4-(cyclohexylamino)benzoate

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₂
MW: 262.35 g/mol
MDL Number: MFCD08072959
CAS Number: 347174-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR