3-Amino-4-chloro-N-(tetrahydro-2-furanylmethyl)benzenesulfonamide

Supplier: Matrix Scientific

053223-500MG 053223-2.5G
101902-780EA 364.62 USD
101902-780 101902-782
3-Amino-4-chloro-N-(tetrahydro-2-furanylmethyl)benzenesulfonamide
3-Amino-4-chloro-N-(tetrahydro-2-furanylmethyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₃S
MW: 290.77 g/mol
MDL Number: MFCD11117892
CAS Number: 1036551-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR