N-(Tetrahydro-2-furanylmethyl)-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053220-500MG
053220-2.5G
101902-768EA
364.62
USD
101902-768
101902-770
N-(Tetrahydro-2-furanylmethyl)-3-piperidinecarboxamide hydrochloride
N-(Tetrahydro-2-furanylmethyl)-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₁ClN₂O₂ |
MDL Number:
MFCD13562642 CAS Number: 1220037-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...