N-(Tetrahydro-2-furanylmethyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053219-500MG 053219-2.5G
101902-764EA 364.62 USD
101902-764 101902-766
N-(Tetrahydro-2-furanylmethyl)-2-piperidinecarboxamide hydrochloride
N-(Tetrahydro-2-furanylmethyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₁ClN₂O₂
MDL Number: MFCD13562641
CAS Number: 1236255-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR