N-(Tetrahydro-2-furanylmethyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053218-500MG
053218-2.5G
101902-760EA
364.62
USD
101902-760
101902-762
N-(Tetrahydro-2-furanylmethyl)-2-pyrrolidinecarboxamide hydrochloride
N-(Tetrahydro-2-furanylmethyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₉ClN₂O₂ |
MDL Number:
MFCD13562640 CAS Number: 1236254-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...