2-Amino-3-phenyl-N-(tetrahydro-2-furanylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

053217-500MG 053217-2.5G
101902-756EA 364.62 USD
101902-756 101902-758
2-Amino-3-phenyl-N-(tetrahydro-2-furanylmethyl)propanamide hydrochloride
2-Amino-3-phenyl-N-(tetrahydro-2-furanylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O₂
MDL Number: MFCD13562639
CAS Number: 1236267-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR