2-Amino-3-methyl-N-(tetrahydro-2-furanylmethyl)butanamide hydrochloride
Supplier: Matrix Scientific
053215-500MG
053215-2.5G
101902-748EA
364.62
USD
101902-748
101902-750
2-Amino-3-methyl-N-(tetrahydro-2-furanylmethyl)butanamide hydrochloride
2-Amino-3-methyl-N-(tetrahydro-2-furanylmethyl)butanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O₂ |
MDL Number:
MFCD13562637 CAS Number: 1236261-04-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...