3-Amino-N-(tetrahydro-2-furanylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053214-500MG
053214-2.5G
101902-744EA
364.62
USD
101902-744
101902-746
3-Amino-N-(tetrahydro-2-furanylmethyl)propanamide hydrochloride
3-Amino-N-(tetrahydro-2-furanylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O₂ |
MDL Number:
MFCD13562636 CAS Number: 1220030-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...