2-(Methylamino)-N-(tetrahydro-2-furanylmethyl)acetamide hydrochloride
Supplier: Matrix Scientific
053213-500MG
053213-2.5G
101902-740EA
364.62
USD
101902-740
101902-742
2-(Methylamino)-N-(tetrahydro-2-furanylmethyl)acetamide hydrochloride
2-(Methylamino)-N-(tetrahydro-2-furanylmethyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O₂ |
MDL Number:
MFCD13562635 CAS Number: 301840-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...