2-Amino-N-(tetrahydro-2-furanylmethyl)propanamide hydrochloride
Supplier: Matrix Scientific
053212-500MG
053212-2.5G
101902-736EA
166.15
USD
101902-736
101902-738
2-Amino-N-(tetrahydro-2-furanylmethyl)propanamide hydrochloride
2-Amino-N-(tetrahydro-2-furanylmethyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O₂ |
MDL Number:
MFCD13562634 CAS Number: 1236266-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...