2-Amino-N-(tetrahydro-2-furanylmethyl)acetamide hydrochloride
Supplier: Matrix Scientific
053211-500MG
053211-2.5G
101902-732EA
364.62
USD
101902-732
101902-734
2-Amino-N-(tetrahydro-2-furanylmethyl)acetamide hydrochloride
2-Amino-N-(tetrahydro-2-furanylmethyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₅ClN₂O₂ |
MDL Number:
MFCD13562633 CAS Number: 890023-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...