6-Chloro-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053210-500MG 053210-2.5G
101902-728EA 364.62 USD
101902-728 101902-730
6-Chloro-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine
6-Chloro-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O
MDL Number: MFCD13562632
CAS Number: 1220035-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR