5-Bromo-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053209-500MG 053209-2.5G
101902-724EA 364.62 USD
101902-724 101902-726
5-Bromo-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine
5-Bromo-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂O
MDL Number: MFCD11119351
CAS Number: 1036500-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR