2-Chloro-N-(tetrahydro-2-furanylmethyl)nicotinamide

Supplier: Matrix Scientific

053207-500MG 053207-2.5G
101902-716EA 364.62 USD
101902-716 101902-718
2-Chloro-N-(tetrahydro-2-furanylmethyl)nicotinamide
2-Chloro-N-(tetrahydro-2-furanylmethyl)nicotinamide

Hazard: Irritant

Formula: C₁₁H₁₃ClN₂O₂
MW: 240.69 g/mol
MDL Number: MFCD03384779
CAS Number: 560102-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR