N2-(Tetrahydro-2-furanylmethyl)-2,3-pyridinediamine
Supplier: Matrix Scientific
053206-500MG
053206-2.5G
101902-712EA
364.62
USD
101902-712
101902-714
N2-(Tetrahydro-2-furanylmethyl)-2,3-pyridinediamine
N2-(Tetrahydro-2-furanylmethyl)-2,3-pyridinediamine
Hazard: Irritant
Formula:
C₁₀H₁₅N₃O |
MDL Number:
MFCD11101071 CAS Number: 1040063-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...