2-[(Tetrahydro-2-furanylmethyl)amino]nicotinic acid
Supplier: Matrix Scientific
053205-500MG
053205-2.5G
101902-708EA
364.62
USD
101902-708
101902-710
2-[(Tetrahydro-2-furanylmethyl)amino]nicotinic acid
2-[(Tetrahydro-2-furanylmethyl)amino]nicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₃ |
MDL Number:
MFCD07757477 CAS Number: 896637-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...