2-[(Tetrahydro-2-furanylmethyl)amino]isonicotinic acid
Supplier: Matrix Scientific
053204-500MG
053204-2.5G
101902-704EA
384.62
USD
101902-704
101902-706
2-[(Tetrahydro-2-furanylmethyl)amino]isonicotinic acid
2-[(Tetrahydro-2-furanylmethyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O₃ |
MDL Number:
MFCD11132188 CAS Number: 1019452-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...