3-Chloro-N-(tetrahydro-2-furanylmethyl)-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

053202-500MG 053202-2.5G
101902-696EA 364.62 USD
101902-696 101902-698
3-Chloro-N-(tetrahydro-2-furanylmethyl)-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-(tetrahydro-2-furanylmethyl)-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₂ClF₃N₂O
MDL Number: MFCD11118279
CAS Number: 1036444-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR