6-Chloro-N-(tetrahydro-2-furanylmethyl)-4-pyrimidinamine

Supplier: Matrix Scientific

053201-500MG 053201-2.5G
101902-692EA 615.38 USD
101902-692 101902-694
6-Chloro-N-(tetrahydro-2-furanylmethyl)-4-pyrimidinamine
6-Chloro-N-(tetrahydro-2-furanylmethyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₉H₁₂ClN₃O
MW: 213.67 g/mol
MDL Number: MFCD13562629
CAS Number: 872512-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR