5-Bromo-4-methyl-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine
Supplier: Matrix Scientific
053200-500MG
053200-2.5G
101902-688EA
364.62
USD
101902-688
101902-690
5-Bromo-4-methyl-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine
5-Bromo-4-methyl-N-(tetrahydro-2-furanylmethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₁H₁₅BrN₂O |
MDL Number:
MFCD13562628 CAS Number: 1220017-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...