Ethyl-3-amino-4-[(tetrahydro-2-furanylmethyl)amino]benzoate
Supplier: Matrix Scientific
053198-500MG
053198-2.5G
101902-680EA
364.62
USD
101902-680
101902-682
Ethyl-3-amino-4-[(tetrahydro-2-furanylmethyl)amino]benzoate
Ethyl-3-amino-4-[(tetrahydro-2-furanylmethyl)amino]benzoate
Hazard: Irritant
Formula:
C₁₄H₂₀N₂O₃ |
MDL Number:
MFCD13344477 CAS Number: 1220016-91-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...