N-(2-Furylmethyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053193-500MG 053193-2.5G
101902-660EA 364.62 USD
101902-660 101902-662
N-(2-Furylmethyl)-2-piperidinecarboxamide hydrochloride
N-(2-Furylmethyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O₂
MW: 244.72 g/mol
MDL Number: MFCD13562624
CAS Number: 205993-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR