2-Amino-N-(2-furylmethyl)-3-phenylpropanamide hydrochloride
Supplier: Matrix Scientific
053191-500MG
053191-2.5G
101902-652EA
364.62
USD
101902-652
101902-654
2-Amino-N-(2-furylmethyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(2-furylmethyl)-3-phenylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₄H₁₇ClN₂O₂ |
MDL Number:
MFCD13562623 CAS Number: 1246172-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...