2-Amino-N-(2-furylmethyl)-2-methylpropanamide hydrochloride

Supplier: Matrix Scientific

053190-500MG 053190-2.5G
101902-648EA 364.62 USD
101902-648 101902-650
2-Amino-N-(2-furylmethyl)-2-methylpropanamide hydrochloride
2-Amino-N-(2-furylmethyl)-2-methylpropanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₅ClN₂O₂
MDL Number: MFCD13562622
CAS Number: 1220035-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR