2-Amino-N-(2-furylmethyl)-3-methylbutanamide hydrochloride
Supplier: Matrix Scientific
053189-500MG
053189-2.5G
101902-644EA
364.62
USD
101902-644
101902-646
2-Amino-N-(2-furylmethyl)-3-methylbutanamide hydrochloride
2-Amino-N-(2-furylmethyl)-3-methylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₇ClN₂O₂ |
MDL Number:
MFCD13562621 CAS Number: 1236256-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...