2-Amino-N-(2-furylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

053186-500MG 053186-2.5G
101902-632EA 364.62 USD
101902-632 101902-634
2-Amino-N-(2-furylmethyl)propanamide hydrochloride
2-Amino-N-(2-furylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂O₂
MDL Number: MFCD13562619
CAS Number: 1236256-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR