2-Amino-N-(2-furylmethyl)acetamide hydrochloride
Supplier: Matrix Scientific
053185-500MG
053185-2.5G
101902-628EA
364.62
USD
101902-628
101902-630
2-Amino-N-(2-furylmethyl)acetamide hydrochloride
2-Amino-N-(2-furylmethyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₇H₁₁ClN₂O₂ |
MDL Number:
MFCD11858019 CAS Number: 1181458-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...